Self-Adhesive Bags with Indicator
Leverancier: MEGRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
355030 355015 355035 355020
MEGR355030EA 54.2 EUR
MEGR355030 MEGR355015 MEGR355035 MEGR355020
Self-Adhesive Bags with Indicator
Zakken Sterilisatiezakken
Steri-bag without fold.
  • Self-adhesive transparent bags
  • Manual closable self-adhesive strip (19 mm wide)
  • With access opening for easy opening by peeling
With steam and EO-gas-indicator.
Order Now
Specificaties
Environmentally Preferable