Rhodium ≥99.8% (metals basis), foil 0.25 mm (0.01 in) thick
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11522.FF 11522.FI
11522.FFEA 0 EUR
11522.FF 11522.FI
Rhodium ≥99.8% (metals basis), foil 0.25 mm (0.01 in) thick
Rhodium
Formule: Rh
MW: 102.9 g/mol
Kookpunt: 3400…3700 °C (1013 hPa)
Smeltpunt: 1960
Dichtheid: 12.41 g/cm³ (20 °C)
MDL Nummer: MFCD00011201
CAS nummer: 7440-16-6
EINECS: 231-125-0
Merck Index: 12,08353