Molecular sieve A4 (0.4 nm, 4 Å), beads ~ 2 mm / ~ 10 mesh, with indicator, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
M-Clarity™ quality level = MQ200
1.05739.0250 1.05739.1000
1.05739.0250EA 110 EUR
1.05739.0250 1.05739.1000
Molecular sieve A4 (0.4 nm, 4 Å), beads ~ 2 mm / ~ 10 mesh, with indicator, Supelco®
Molecular sieve A4 (0.4 nm, 4 Å)
Dichtheid: 1,1…2.2 g/cm³ (20 °C)
MDL Nummer: MFCD03457537
CAS nummer: 70955-01-0