tert-Butyl-4-methoxyphenol (mixture of isomers) 96%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: tert-Butyl-4-methoxyphenol, BHA, tert-Butyl-4-hydroxyanisole, Butylhydroxyanisole, 2(3)-tert-Butyl-4-methoxyphenol, Butylated hydroxyanisole
235231000 235235000 235230025
ACRO235231000EA 36.1 EUR
ACRO235231000 ACRO235235000 ACRO235230025
tert-Butyl-4-methoxyphenol (mixture of isomers) 96%
tert-Butyl-4-methoxyphenol (mixture of isomers)
Formule: C₁₁H₁₆O₂
Kookpunt: 268 °C (760 mmHg)
Smeltpunt: 48…55 °C
Dichtheid: 1,05 g/cm³ (20 °C)
Vlampunt: 113 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01779059
CAS nummer: 25013-16-5
EINECS: 246-563-8
Merck Index: 12,01582
Order Now
Specification Test Results
Appearance White to light yellow Crystals or flakes
Infrared spectrum Conforms
GC ≥95 %