tert-Butyl-4-methoxyphenol (mixture of isomers) ≥96%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: tert-Butyl-4-methoxyphenol, BHA, tert-Butyl-4-hydroxyanisole, Butylhydroxyanisole, 2(3)-tert-Butyl-4-methoxyphenol, Butylated hydroxyanisole
B23530.0B B23530.18 B23530.30
B23530.0BEA 258 EUR
B23530.0B B23530.18 B23530.30
tert-Butyl-4-methoxyphenol (mixture of isomers) ≥96%
tert-Butyl-4-methoxyphenol (mixture of isomers)
Purity: 96% CAS No.: 25013-16-5 Molecular Formula: C11H16O2 Formula Weight: 180.25 Melting Point: 55-65° Merck Reference: 14,1547 MDL No.: MFCD01779059
Formule: C₁₁H₁₆O₂
Kookpunt: 264 °C (1013 hPa)
Smeltpunt: 55…65 °C
Dichtheid: 1,05 g/cm³ (20 °C)
Vlampunt: 156 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01779059
CAS nummer: 25013-16-5
EINECS: 246-563-8
Merck Index: 14,1547