Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture, anhydrous 99%, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Decalin, Bicyclo[4.4.0]decane, Decahydronaphthalene
406171000 406170010
406171000.EA 42.7 EUR
406171000. 406170010.
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture, anhydrous 99%, AcroSeal®
Decahydronaphthalene (mixture of cis and trans isomers)
Formule: C₁₀H₁₈
MW: 138.25 g/mol
Kookpunt: 187 °C (760 mmHg)
Smeltpunt: –31 °C
Dichtheid: 0.89 g/cm³
Vlampunt: 58 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004130
CAS nummer: 91-17-8
EINECS: 202-046-9
UN: 1147
ADR: 3 III
Merck Index: 13,02866
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41b5fff8aa81be0e42af177a8f0e3ac149943c6e&vid=ca9f3c5d71ac621a6216f8b8b28d7cb27b29fd84&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.5 % (mixture of cis and trans)