1,4-Cyclohexanediol cis- and trans-mixture ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Cyclohexane-1,4-diol
B20446.14 B20446.22
B20446.14EA 28.5 EUR
B20446.14 B20446.22
1,4-Cyclohexanediol cis- and trans-mixture ≥98%
1,4-Cyclohexanediol
Purity: 98% CAS No.: 556-48-9 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 65°(149°F) Melting Point: 98-103° Boiling Point: 149-151°/20mm Beilstein Reference: 2036658 MDL No.: MFCD00001448
Formule: C₆H₁₂O₂
MW: 116.16 g/mol
Kookpunt: 149…151 °C (20 mmHg)
Smeltpunt: 98…103 °C
Dichtheid: 1.17 g/cm³ (21.85 °C)
Vlampunt: 65 °C (149 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001448
CAS nummer: 556-48-9
EINECS: 209-126-2