Paraffin, Solidification point: 42-44°C
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Wax Paraffin, Paraffin wax
M-Clarity™ quality level = MQ200
1.07150.1000 1.07150.9025 1.07150.2500
1.07150.1000EA 154 EUR
1.07150.1000 1.07150.9025 1.07150.2500
Paraffin, Solidification point: 42-44°C
Paraffin
Dichtheid: 0,9 g/cm³ (20 °C)
CAS nummer: 8002-74-2
EINECS: 232-315-6