Molecular sieve 13X (1.0 nm, 10 Å)
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Linde 13X
8483.1 8483.2
ROTH8483.1EA 0 EUR
ROTH8483.1 ROTH8483.2
Molecular sieve 13X (1.0 nm, 10 Å)
Molecular sieve 13X (1.0 nm, 10 Å)
MDL Nummer: MFCD00164506
CAS nummer: 63231-69-6