Molecular sieve A4 (0.4 nm, 4 Å)
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
8471.1 8471.2
ROTH8471.1EA 0 EUR
ROTH8471.1 ROTH8471.2
Molecular sieve A4 (0.4 nm, 4 Å)
Molecular sieve A4 (0.4 nm, 4 Å)
Dichtheid: 1,1…2.2 g/cm³ (20 °C)
MDL Nummer: MFCD03457537
CAS nummer: 70955-01-0