Molecular sieve A5 (0.5 nm, 5 Å), pellets, Ø 3-4mm (0.12-0.16in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets
33551.A1 33551.A7
33551.A1EA 110 EUR
33551.A1 33551.A7
Molecular sieve A5 (0.5 nm, 5 Å), pellets, Ø 3-4mm (0.12-0.16in)
Molecular sieve A5 (0.5 nm, 5 Å)
MDL Nummer: MFCD00131613
CAS nummer: 69912-79-4