Petroleum spirit, 35…60 °C ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

42085.K2 42085.M1
42085.K2EA 59.2 EUR
42085.K2 42085.M1
Petroleum spirit, 35…60 °C ACS
Petroleum spirit

CAS No.: 8032-32-4 Flash Point: -49°(-57°F) Physical Form: Liquid Density: 0.640 Refractive Index: 1.3630 Appearance: Colourless Odour: Petroleum-like Boiling Point: 35-60° UN No.: 1268 MDL No.: MFCD00081849

Kookpunt: 35…60 °C
Dichtheid: 0,640
Vlampunt: –49 °C (–57 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081849
CAS nummer: 64742-49-0
EINECS: 232-453-7
UN: 1268
ADR: 3,I

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR