Molecular sieve A5 (0.5 nm, 5 Å), beads 8-12 mesh, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets
208620-1KG 208620-5KG
208620-1KGEA 221 EUR
208620-1KG 208620-5KG
Molecular sieve A5 (0.5 nm, 5 Å), beads 8-12 mesh, Fluka™
Molecular sieve A5 (0.5 nm, 5 Å)
MDL Nummer: MFCD00131613
CAS nummer: 69912-79-4
Order Now
Specification Test Results
Form beads
Grain size 8 - 12 mesh Min. 95 %