Petroleum spirit, 40…65 °C, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Gasoline, Petroleum ether, Petrol, Benzine
8115.5000EA 266.19 EUR
8115.5000 8115.1000 8115.9025 8115.9200
Petroleum spirit, 40…65 °C, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Petroleum spirit
Kookpunt: 40…65 °C (1013 hPa)
Dichtheid: 0.65 g/cm³ (20 °C)
Vlampunt: –40 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081849
CAS nummer: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II
Order Now
Specification Test Results
Boiling Range 40 - 65 °C
Residue after Evaporation max. 0.001 %
Water (H2O) max. 0.02 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1