Petroleum spirit, 40…60 °C

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Gasoline, Petroleum ether, Petrol, Benzine

9320.1 9320.2
ROTH9320.1EA 0 EUR
ROTH9320.1 ROTH9320.2
Petroleum spirit, 40…60 °C
Petroleum spirit
Kookpunt: 40…60 °C (1013 hPa)
Dichtheid: 0,65 g/cm³ (20 °C)
Vlampunt: –40 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081849
CAS nummer: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR