Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Gasoline, Petroleum ether, Petrol, Benzine
23837.553-200
23837.325EA 45.6 EUR
23837.325 23837.291 23837.360 23837.462 23837.553 XXXX23837.553-200
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Petroleum spirit
Kookpunt: 40…65 °C (1013 hPa)
Dichtheid: 0.65 g/cm³ (20 °C)
Vlampunt: –40 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081849
CAS nummer: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II
Order Now
Specification Test Results
Distillation range 40 - 65 °C