Dimethyl sulphoxide-[D₆] 99.8 atom % D + 0.03% (v/v) TMS NMR
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6
360000100 360000500
ACRO360000100EA 62.6 EUR
ACRO360000100 ACRO360000500
Dimethyl sulphoxide-[D₆] 99.8 atom % D + 0.03% (v/v) TMS NMR
Dimethyl sulphoxide-[D₆]
Formule: D₃CS(O)CD₃
MW: 84.09 g/mol
Kookpunt: 189
Smeltpunt: 20.2
Dichtheid: 1.19 g/cm³
Vlampunt: 88 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002090
CAS nummer: 2206-27-1
EINECS: 218-617-0
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.03 % (Coulometric)
D-Enrichment ≥99.75 %
TMS 0.02 to 0.04 v/v %