Quaternaire ammoniumverbindingen, benzyl-C8-18-alkyldimethyl, chloriden
Leverancier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
14520.02 14520.03
SERA14520.02EA 0 EUR
SERA14520.02 SERA14520.03
Quaternaire ammoniumverbindingen, benzyl-C8-18-alkyldimethyl, chloriden
Quaternaire ammoniumverbindingen, benzyl-C8-18-alkyldimethyl, chloriden
Formule: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
Smeltpunt: 29…34 °C
Dichtheid: 0.98 g/cm³ (20 °C)
MDL Nummer: MFCD00145757
CAS nummer: 63449-41-2
EINECS: 264-151-6
UN: 2923
ADR: 8 III