D-(+)-Maltose monohydrate 92+%
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ACRO329915000EA 120 EUR
ACRO329915000 ACRO329911000 ACRO329910025
D-(+)-Maltose monohydrate 92+%
D-(+)-Maltose monohydrate
Formule: C₁₂H₂₂O₁₁·1H₂O
MW: 360,32 g/mol
Smeltpunt: 130 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149343
CAS nummer: 6363-53-7
EINECS: 200-716-5
Merck Index: 13,05736
https://nl.vwr.com/stibo/web/std.lang.all/21/24/2302124.jpg
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤7 %
Sulfated ash ≤0.1 %
Additional info remainder mainly glucose and maltotriose
HPLC ≥92.0 %
Specific optical rotation +135° to +139° (20°C, 589 nm) (c=4, H2O, NH3) (on dry substance)