D-(-)-Sorbitol FP, liquid, EMPROVE® ESSENTIAL E420, BP, Ph. Eur., NF suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
1.02975.9050 1.02975.9270
1.02975.9050EA 942 EUR
1.02975.9050 1.02975.9270
D-(-)-Sorbitol FP, liquid, EMPROVE® ESSENTIAL E420, BP, Ph. Eur., NF suitable for use as excipient, SAFC®
D-(-)-Sorbitol
Sorbitol FP liquid (sorbitol syrup, non crystallizing)
Formule: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Kookpunt: 295
Dichtheid: 1.3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004708
CAS nummer: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM