Sucrose (saccharose), EMPROVE® EXPERT BP, Ph. Eur., JP, NF for density gradient ultracentrifugation, for biopharmaceutical production, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
1.07654.1000 1.07654.5000 1.07654.9029
1.07654.1000EA 466 EUR
1.07654.1000 1.07654.5000 1.07654.9029
Sucrose (saccharose), EMPROVE® EXPERT BP, Ph. Eur., JP, NF for density gradient ultracentrifugation, for biopharmaceutical production, SAFC®
D-(+)-Sucrose
Formule: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Smeltpunt: 169…170 °C
Dichtheid: 1.29 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006626
CAS nummer: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM