D-(+)-Sucrose

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose

35579.01 35579.02 35579.03 35579.04
SERA35579.01EA 0 EUR
SERA35579.01 SERA35579.02 SERA35579.03 SERA35579.04
D-(+)-Sucrose
D-(+)-Sucrose
Formule: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Smeltpunt: 169…170 °C
Dichtheid: 1.29 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006626
CAS nummer: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR