Agarose
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11382.01
11382.02
11382.03
SERA11382.01EA
0
EUR
SERA11382.01
SERA11382.02
SERA11382.03
Agarose
Agarose
Smeltpunt:
60…90 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00081294 CAS nummer: 9012-36-6 EINECS: 232-731-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...