Safety storage cabinets, CX-Classic-G
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
32925041 32925050
MSGA32925041EA 5040 EUR
MSGA32925041 MSGA32925050
Safety storage cabinets, CX-Classic-G
Meubilair Opslagkasten
Fireproof safety cabinets for safe and approved storage of water pollutants, non flammable liquids and toxics in working areas according to TRGS 510.
  • Robust construction and longevity- high-quality, scratch-resistant outer body with powder-coated textured surface
  • Doors lockable with cylinder locking (integrated in the turning handle)
  • Adjusting aids to compensate for uneven floor
Cabinet body colour: anthracite grey RAL 7016.
Cabinet features with perforated insert useable as storage area. It consists of bottom collecting sump with galvanised or painted version, liquid-tight
Recirculating air filter system is ideal for the direct installation in working areas with very low noise, only approximately 39 dB. It consists of multi-stage wideband filter, cool-device cable, optical and acoustical alarm including potential-free alarm contact, 4,7" display with innovative touch panel and manual check of the filter saturation
Certificaten: Complies with VDE approval of the monitoring electronics.
Bestelinformatie: Interior equipment packages and interior equipments should be ordered separately.