Zetmeel, hydrolysed acc. Smithies

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

35370.01 35370.02 35370.03
SERA35370.01EA 0 EUR
SERA35370.01 SERA35370.02 SERA35370.03
Zetmeel, hydrolysed acc. Smithies
Zetmeel
Formule: (C₆H₁₀O₅)n
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00082026
CAS nummer: 9005-25-8
EINECS: 232-679-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR