Gold germanium alloy (88:12 w%) ≥99.998% (metals basis), slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42983.KJ
42983.KM
42983.KJEA
572
EUR
42983.KJ
42983.KM
Gold germanium alloy (88:12 w%) ≥99.998% (metals basis), slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Gold germanium alloy (88:12 w%)
Purity: 99.998%
Molecular Formula: Au:Ge; 88:12 wt%
Physical Form: ≈0.4g/slug
Density: 0.37G/CM(3)
Odour: Odourless
MDL No.: MFCD02091706