Copper nickel alloy (55:45 w%), draad, Ø 0.075 mm (0.003 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43269.H4
43269.H9
43269.H4EA
40.5
EUR
43269.H4
43269.H9
Copper nickel alloy (55:45 w%), draad, Ø 0.075 mm (0.003 in)
Copper nickel alloy (55:45 w%)
Molecular Formula: Cu:Ni 55:45 wt%
Physical Form: ≈0.04g/m
MDL No.: MFCD00801098
Resistance alloy, negative lead for type J thermocouple