Copper nickel alloy (55:45 w%), wire, Ø 0.13 mm (0.005 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
43270.H4
43270.H9
43270.H4EA
37.2
EUR
43270.H4
43270.H9
Copper nickel alloy (55:45 w%), wire, Ø 0.13 mm (0.005 in)
Copper nickel alloy (55:45 w%)
Molecular Formula: Cu:Ni 55:45 wt%
Physical Form: ≈0.11g/m
Odour: Odourless
MDL No.: MFCD00801098