Palladium silver alloy (75:25 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness: 0.25 mm (0.01 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42683.FF
42683.FI
42683.FFEA
452
EUR
42683.FF
42683.FI
Palladium silver alloy (75:25 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness: 0.25 mm (0.01 in)
Palladium silver alloy (75:25 w%)
Purity: 99.9%
Molecular Formula: Pd:Ag; 75:25 wt%
Physical Form: ≈1.82g/25x25mm
Odour: Odourless
MDL No.: MFCD02091738