Palladium silver alloy (75:25 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness: 0.5 mm (0.02 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42684.FF
42684.FI
42684.FFEA
901
EUR
42684.FF
42684.FI
Palladium silver alloy (75:25 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness: 0.5 mm (0.02 in)
Palladium silver alloy (75:25 w%)
Purity: 99.9%
Molecular Formula: Pd:Ag; 75:25 wt%
Physical Form: ≈3.64g/25x25mm
Odour: Odourless
MDL No.: MFCD02091738