Palladium silver alloy (77:23 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness: 0.025 mm (0.001 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45003.FI
45003.FF
45003.FIEA
549
EUR
45003.FI
45003.FF
Palladium silver alloy (77:23 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness: 0.025 mm (0.001 in)
Palladium silver alloy (77:23 w%)
Purity: 99.9%
Molecular Formula: Pd:Ag; 77:23 wt%
Physical Form: ≈0.18g/25x25mm
Odour: Odourless
MDL No.: MFCD02091738