Platinum rhodium alloy (70:30 w%), wire for ISA type B standard grade thermocouple, Ø 0.25 mm (0.01 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10063.G1
10063.BY
10063.G1EA
0
EUR
10063.G1
10063.BY
Platinum rhodium alloy (70:30 w%), wire for ISA type B standard grade thermocouple, Ø 0.25 mm (0.01 in)
Platinum rhodium alloy (70:30 w%)
Molecular Formula: Pt:Rh; 70:30 wt%
Physical Form: ≈0.52g/50cm
Odour: Odourless
MDL No.: MFCD00198186