Platinum iridium alloy (80:20 w%), wire, hard, Ø 0.5 mm (0.02 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10054.BS
10054.BW
10054.BSEA
136
EUR
10054.BS
10054.BW
Platinum iridium alloy (80:20 w%), wire, hard, Ø 0.5 mm (0.02 in)
Platinum iridium alloy (80:20 w%)
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈0.44g/10cm, Temper: as drawn
Odour: Odourless
MDL No.: MFCD02091683