Platinum iridium alloy (80:20 w%) 99,9 Metals basis exPreciousMetals, wire, hard, Ø 0.127 mm (0.005 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43330.BS
43330.BW
43330.BSEA
56.8
EUR
43330.BS
43330.BW
Platinum iridium alloy (80:20 w%) 99,9 Metals basis exPreciousMetals, wire, hard, Ø 0.127 mm (0.005 in)
Platinum iridium alloy (80:20 w%)
Purity: 99.9%
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈0.03g/10cm, Temper: hard
Odour: Odourless
MDL No.: MFCD02091683