Platinum rhodium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41811.FI
41811.GH
41811.FIEA
2610
EUR
41811.FI
41811.GH
Platinum rhodium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Platinum rhodium alloy (80:20 w%)
Purity: 99.9%
Molecular Formula: Pt:Rh; 80:20 wt%
Physical Form: ≈0.29g/25x25mm
Odour: Odourless
MDL No.: MFCD00198186