Platinum rhodium alloy (80:20 w%) ≥99.9% (metals basis), draad, Ø 0.25 mm (0.01 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43274.BY
43274.BU
43274.BYEA
621
EUR
43274.BY
43274.BU
Platinum rhodium alloy (80:20 w%) ≥99.9% (metals basis), draad, Ø 0.25 mm (0.01 in)
Platinum rhodium alloy (80:20 w%)
Purity: 99.9%
Molecular Formula: Pt:Rh; 80:20 wt%
Physical Form: ≈0.24g/25cm
Odour: Odourless
MDL No.: MFCD00198186