Platinum iridium alloy (85:15 w%), draad, annealed, Ø 0.25 mm (0.01 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43275.BY
43275.BU
43275.BYEA
805
EUR
43275.BY
43275.BU
Platinum iridium alloy (85:15 w%), draad, annealed, Ø 0.25 mm (0.01 in)
Platinum iridium alloy (85:15 w%)
Molecular Formula: Pt:Ir; 85:15 wt%
Physical Form: ≈0.27g/25cm, Temper: soft
Odour: Odourless
MDL No.: MFCD02091683