Platinum rhodium alloy (87:13 w%), draad for ISA type R standard grade thermocouple, Ø 0.25 mm (0.01 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10062.BU
10062.BY
10062.CC
10062.BUEA
174
EUR
10062.BU
10062.BY
10062.CC
Platinum rhodium alloy (87:13 w%), draad for ISA type R standard grade thermocouple, Ø 0.25 mm (0.01 in)
Platinum rhodium alloy (87:13 w%)
Molecular Formula: Pt:Rh; 87:13 wt%
Physical Form: ≈0.26g/25cm
Odour: Odourless
MDL No.: MFCD00198186