Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.076 mm (0.003 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42902.BW
42902.G2
42902.G6
42902.BWEA
84.4
EUR
42902.BW
42902.G2
42902.G6
Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.076 mm (0.003 in)
Platinum rhodium alloy (87:13 w%)
Molecular Formula: Pt:Rh; 87:13 wt%
Physical Form: ≈0.045g/50cm
Odour: Odourless
MDL No.: MFCD00198186