Platinum rhodium alloy (87:13 w%), wire thermocouple_ISA_type_R, Ø 0.203 mm (0.008 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42905.G1
42905.G5
42905.G9
42905.G1EA
443
EUR
42905.G1
42905.G5
42905.G9
Platinum rhodium alloy (87:13 w%), wire thermocouple_ISA_type_R, Ø 0.203 mm (0.008 in)
Platinum rhodium alloy (87:13 w%)
Molecular Formula: Pt:Rh; 87:13 wt%
Physical Form: ≈0.64g/m
Odour: Odourless
MDL No.: MFCD00198186