Platinum iridium alloy (90:10 w%) 99,9 Metals basis, wire, annealed, Ø 0.25 mm (0.01 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
39383.BS
39383.BW
39383.BSEA
94.7
EUR
39383.BS
39383.BW
Platinum iridium alloy (90:10 w%) 99,9 Metals basis, wire, annealed, Ø 0.25 mm (0.01 in)
Platinum iridium alloy (90:10 w%)
Purity: 99.9+%
Molecular Formula: Pt:Ir; 90:10 wt%,
Physical Form: ≈0.11g/10cm, Temper: soft
Odour: Odourless
MDL No.: MFCD02091683