Platinum iridium alloy (90:10 w%), gauze woven from wire 150 mesh, Ø 0.043 mm (0.0017 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40934.FF
40934.FI
40934.GH
40934.FFEA
275
EUR
40934.FF
40934.FI
40934.GH
Platinum iridium alloy (90:10 w%), gauze woven from wire 150 mesh, Ø 0.043 mm (0.0017 in)
Platinum iridium alloy (90:10 w%)
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: Wire Cloth
Odour: Odourless
MDL No.: MFCD02091683
Filtration, electrodes