Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis excluding precious metals), foil, Thickness: 0.05 mm (0.002 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41803.FF
41803.FI
41803.GH
41803.FFEA
272
EUR
41803.FF
41803.FI
41803.GH
Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis excluding precious metals), foil, Thickness: 0.05 mm (0.002 in)
Platinum iridium alloy (90:10 w%)
Purity: 99.9%
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈0.67g/25x25mm
Odour: Odourless
MDL No.: MFCD02091683