Platinum iridium alloy (90:10 w%) 99,9 Metals basis, foil, Thickness: 0.1 mm (0.004 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41804.FF
41804.FI
41804.GH
41804.FFEA
364
EUR
41804.FF
41804.FI
41804.GH
Platinum iridium alloy (90:10 w%) 99,9 Metals basis, foil, Thickness: 0.1 mm (0.004 in)
Platinum iridium alloy (90:10 w%)
Purity: 99.9%
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈1.35g/25x25mm
Odour: Odourless
MDL No.: MFCD02091683