Platinum iridium alloy (90:10 w%), wire, Ø 1.0 mm (0.04 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42678.BQ
42678.BU
42678.BY
42678.BQEA
600
EUR
42678.BQ
42678.BU
42678.BY
Platinum iridium alloy (90:10 w%), wire, Ø 1.0 mm (0.04 in)
Platinum iridium alloy (90:10 w%)
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈0.85g/5cm
Odour: Odourless
MDL No.: MFCD02091683