Platinum iridium alloy (90:10 w%), wire, Ø 1.5 mm (0.06 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
44812.BQ
44812.BU
44812.BQEA
1290
EUR
44812.BQ
44812.BU
Platinum iridium alloy (90:10 w%), wire, Ø 1.5 mm (0.06 in)
Platinum iridium alloy (90:10 w%)
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈1.91g/5cm
Odour: Odourless
MDL No.: MFCD02091683