Platinum rhodium alloy (90:10 w%) 99,9 Metals basis, foil, Thickness: 0.025 mm (0.001 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41808.FI
41808.GH
41808.FIEA
2764
EUR
41808.FI
41808.GH
Platinum rhodium alloy (90:10 w%) 99,9 Metals basis, foil, Thickness: 0.025 mm (0.001 in)
Platinum rhodium alloy (90:10 w%)
Purity: 99.9%
Molecular Formula: Pt:Rh; 90:10 wt%
Physical Form: ≈0.31g/25x25mm
Odour: Odourless
MDL No.: MFCD00198186