Platinum rhodium alloy (90:10 w%), wire for ISA type S standard grade thermocouple, Ø 0.127 mm (0.005 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42893.G1
42893.G5
42893.G1EA
337
EUR
42893.G1
42893.G5
Platinum rhodium alloy (90:10 w%), wire for ISA type S standard grade thermocouple, Ø 0.127 mm (0.005 in)
Platinum rhodium alloy (90:10 w%)
Molecular Formula: Pt:Rh; 90:10 wt%
Physical Form: ≈0.25g/m
Odour: Odourless
MDL No.: MFCD00198186