Platinum rhodium alloy (90:10 w%), wire for ISA type S standard grade thermocouple, Ø 0.203 mm (0.008 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42894.G1
42894.G5
42894.G9
42894.G1EA
435
EUR
42894.G1
42894.G5
42894.G9
Platinum rhodium alloy (90:10 w%), wire for ISA type S standard grade thermocouple, Ø 0.203 mm (0.008 in)
Platinum rhodium alloy (90:10 w%)
Molecular Formula: Pt:Rh; 90:10 wt%
Physical Form: ≈0.65g/m
Odour: Odourless
MDL No.: MFCD00198186