Platinum rhodium alloy (94:6 w%), wire for ISA type B standard grade thermocouple, Ø 0.5 mm (0.02 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42914.BU
42914.BY
42914.BUEA
667
EUR
42914.BU
42914.BY
Platinum rhodium alloy (94:6 w%), wire for ISA type B standard grade thermocouple, Ø 0.5 mm (0.02 in)
Platinum rhodium alloy (94:6 w%)
Molecular Formula: Pt:Rh; 94:6 wt%
Physical Form: ≈1.04g/25cm
Odour: Odourless
MDL No.: MFCD00198186